1-Bromo-4-nitrobenzene, 98%
1-Bromo-4-nitrobenzene, 98%
1-Bromo-4-nitrobenzene, 98%
Thermo Scientific Chemicals

1-Bromo-4-nitrobenzene, 98%

CAS: 586-78-7 | C6H4BrNO2 | 202.01 g/mol
Quantity:
5 g
25 g
100 g
500 g
Catalog number A17705.22
also known as A17705-22
Price (JPY)
-
Quantity:
100 g
Request bulk or custom format
Chemical Identifiers
CAS586-78-7
IUPAC Name1-bromo-4-nitrobenzene
Molecular FormulaC6H4BrNO2
InChI KeyZDFBKZUDCQQKAC-UHFFFAOYSA-N
SMILES[O-][N+](=O)C1=CC=C(Br)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)≥97.5%
Appearance (Color)White to cream to orange to brown
FormPowder
Identification (FTIR)Conforms
Melting Point (clear melt)122.0-128.0?C
It can be employed as a syntheses material intermediate. It is also used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It can be employed as a syntheses material intermediate. It is also used as a pharmaceutical intermediate.

Solubility
Insoluble in water. Almost transparent in hot ethyl alcohol. Soluble in ether

Notes
Store away from oxidizing agents and bases. Keep the container tightly closed in a cool, dry place.
RUO – Research Use Only

General References:

  1. Sven Ernst.; Sarah E Norman.; Christopher Hardacre.; Richard G Compton. The electrochemical reduction of 1-bromo-4-nitrobenzene at zinc electrodes in a room-temperature ionic liquid: a facile route for the formation of arylzinc compounds.Phys Chem Chem Phys.2014,16(10), 4478-4482.
  2. Jin-Heng Li.; Yun Liang.; De-Ping Wang.; Wen-Jie Liu.; Ye-Xiang Xie.; Du-Lin Yin. Efficient Stille cross-coupling reaction catalyzed by the Pd(OAc)2/Dabco catalytic system.J Org Chem.2005,70(7), 2832-2834.