Thermo Scientific Chemicals

L-Isoleucine methyl ester hydrochloride, 98+%, Thermo Scientific Chemicals

Catalog number: A17847.06
5g, Each
Thermo Scientific Chemicals

L-Isoleucine methyl ester hydrochloride, 98+%, Thermo Scientific Chemicals

Catalog number: A17847.06
5g, Each
Quantity
Catalog number: A17847.06
also known as A17847-06
Price (USD)

Chemical Identifiers

CAS
18598-74-8
IUPAC Name
methyl (2S,3S)-2-amino-3-methylpentanoate hydrochloride
Molecular Formula
C7H16ClNO2
InChI Key
GGTBEWGOPAFTTH-GEMLJDPKSA-N
SMILES
Cl.CC[C@H](C)[C@H](N)C(=O)OC
Optical Rotation
+27? ?1? (c=2 in water)
Appearance (Color)
White
Form
Crystals or powder or crystalline powder
Assay (Titration ex Chloride)
≥98.0 to ≤102.0%
Water Content (Karl Fischer Titration)
≤1.0%

Description

L-Isoleucine methyl ester used for chemical synthesis. E.g. It can also be used to produce 3-methyl-2-(4-nitro-imidazol-1-yl)-pentanoic acid methyl ester. It also finds its use as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
L-Isoleucine methyl ester used for chemical synthesis. E.g. It can also be used to produce 3-methyl-2-(4-nitro-imidazol-1-yl)-pentanoic acid methyl ester. It also finds its use as a pharmaceutical intermediate.

Solubility
Soluble in methanol (50 mg/ml clear, colorless solution).

Notes
Hygroscopic. Keep away from oxidizing agents, water/moisture. Store under inert atmosphere. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. In order to obtain a higher solubility, warm the tube at 37°C and shake it in the ultrasonic bath for a while.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text