Thermo Scientific Chemicals

Ethylphosphonic dichloride, 98%, Thermo Scientific Chemicals

Catalog number: A17869.14
25 g, Each
Thermo Scientific Chemicals

Ethylphosphonic dichloride, 98%, Thermo Scientific Chemicals

Catalog number: A17869.14
25 g, Each
Quantity

Chemical Identifiers

CAS
1066-50-8
IUPAC Name
ethylphosphonoyl dichloride
Molecular Formula
C2H5Cl2OP
InChI Key
OWGJXSYVHQEVHS-UHFFFAOYSA-N
SMILES
CCP(Cl)(Cl)=O
Appearance (Color)
Clear colorless
Refractive Index
1.4645-1.4685 @ 20?C
Form
Liquid
Assay (GC)
≥97.5%

Description

Ethylphosphonic dichloride finds it uses in the preparation of: ethylphosphonic diisocyanate, 4-acetylphenyl isopropyl ethylphosphonate, 4-acetylphenyl phenyl ethylphosphonate and 4-acetylphenyl cyclohexyl ethylphosphonate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ethylphosphonic dichloride finds it uses in the preparation of: ethylphosphonic diisocyanate, 4-acetylphenyl isopropyl ethylphosphonate, 4-acetylphenyl phenyl ethylphosphonate and 4-acetylphenyl cyclohexyl ethylphosphonate.

Solubility
Soluble in chloroform and dichloromethane.

Notes
Moisture Sensitive. Store away from oxidizing agents, water/moisture. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text