meso-2,3-Dimercaptosuccinic acid, 97%
meso-2,3-Dimercaptosuccinic acid, 97%
meso-2,3-Dimercaptosuccinic acid, 97%
meso-2,3-Dimercaptosuccinic acid, 97%
Thermo Scientific Chemicals

meso-2,3-Dimercaptosuccinic acid, 97%

CAS: 304-55-2 | C4H6O4S2 | 182.208 g/mol
Quantity:
1 g
5 g
Catalog number A17909.06
also known as A17909-06
Price (JPY)
-
Request A Quote
Quantity:
5 g
Request bulk or custom format
Chemical Identifiers
CAS304-55-2
IUPAC Name2,3-disulfanylbutanedioic acid
Molecular FormulaC4H6O4S2
InChI KeyACTRVOBWPAIOHC-UHFFFAOYNA-N
SMILESOC(=O)C(S)C(S)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Assay (Aqueous acid-base Titration)≥96.0 to ≤104.0%
Identification (FTIR)Conforms
FormPowder
It is applied as a selective masking agent and chelating agent for cadmium in EDTA titration of zinc.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is applied as a selective masking agent and chelating agent for cadmium in EDTA titration of zinc.

Solubility
Soluble in water, and ethanol (25 mg/ml, results in colorless to light yellow, clear).

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
RUO – Research Use Only

General References:

  1. Vineeta N.; Singh V.; Makkar S. Anti microbial activity of dimercaptosuccinic acid (DMSA): a new chelating agent.J Indian Soc Pedod Prev Dent.,2001,19(4), 160-163.
  2. Kramer HJ.; Mensikova V.; Bäcker A.; Meyer-Lehnert H.; Gonick HC. Interaction of dimercaptosuccinic acid (DMSA) with angiotensin II on calcium mobilization in vascular smooth muscle cells.Biochem Pharmacol.,2003,65(10), 1741-6.
  3. Water soluble chelating agent.