(1-Dodecyl)trimethylammonium chloride, 97%
(1-Dodecyl)trimethylammonium chloride, 97%
(1-Dodecyl)trimethylammonium chloride, 97%
Thermo Scientific Chemicals

(1-Dodecyl)trimethylammonium chloride, 97%

CAS: 112-00-5 | C15H34ClN | 263.894 g/mol
Quantity:
5 g
25 g
100 g
Catalog number A17924.22
also known as A17924-22
Price (JPY)
-
Quantity:
100 g
Request bulk or custom format
Chemical Identifiers
CAS112-00-5
IUPAC Namedodecyltrimethylazanium chloride
Molecular FormulaC15H34ClN
InChI KeyDDXLVDQZPFLQMZ-UHFFFAOYSA-M
SMILES[Cl-].CCCCCCCCCCCC[N+](C)(C)C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to pale yellow
Assay from Suppliers CofA≥96.0%
FormPowder or crystals or crystalline powder
Infrared spectrumConforms
Dodecyltrimethylammonium Chloride is useful as a paint stripper, a foaming stabilizer, and a bactericidal lotion. It is used in coating, plastics, paints, rubber and ink production, as anti-static agent, catalyst.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Dodecyltrimethylammonium Chloride is useful as a paint stripper, a foaming stabilizer, and a bactericidal lotion. It is used in coating, plastics, paints, rubber and ink production, as anti-static agent, catalyst.

Solubility
Soluble in water.

Notes
Hygroscopic. Store under inert gas. Store away from oxidizing agents, moisture.
RUO – Research Use Only

General References:

  1. Ramkrishna Adhikary; Philip J Carlson; Tak W Kee; Jacob W Petrich. Excited-state intramolecular hydrogen atom transfer of curcumin in surfactant micelles. Journal of Physical Chemistry. 2010, 114, (8), 2997-3004.
  2. Jonas Carlstedt; Azat Bilalov; Ulf Olsson. Aqueous phase behavior of polyelectrolytes with amphiphilic counterions modulated by cyclodextrin: the role of polyion flexibility. Physical Chemistry Chemical Physics. 2012, 14, (27), 9574-9577.