Thermo Scientific Chemicals

Di-n-octyl sulfide, 97%, Thermo Scientific Chemicals

Catalog number: A17955.36
500 g, Each
Thermo Scientific Chemicals

Di-n-octyl sulfide, 97%, Thermo Scientific Chemicals

Catalog number: A17955.36
500 g, Each
Quantity
Catalog number: A17955.36
also known as A17955-36
Price (USD)
Price: 410.00
Online price: 348.65
Your price:
Quantity
-

Chemical Identifiers

CAS
2690-08-6
IUPAC Name
1-(octylsulfanyl)octane
Molecular Formula
C16H34S
InChI Key
LOXRGHGHQYWXJK-UHFFFAOYSA-N
SMILES
CCCCCCCCSCCCCCCCC
Form
Liquid
Identification (FTIR)
Conforms
Appearance (Color)
Clear colorless
Assay (GC)
≥96.0%
Refractive Index
1.4595-1.4645 @ 20?C

Description

It is employed as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is employed as a pharmaceutical intermediate.

Solubility
Solubility in water, 0.004678 mg/L @ 25°C (est)

Notes
Air & Moisture Sensitive. Store away from acids, oxidizing agents, air, water/moisture. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text