5-Bromovaleryl chloride, 98%
5-Bromovaleryl chloride, 98%
Thermo Scientific Chemicals

5-Bromovaleryl chloride, 98%

CAS: 4509-90-4 | C14H11N3O2 | 253.26 g/mol
Quantity:
5 g
25 g
Catalog number A18046.14
also known as A18046-14
Price (JPY)
-
Request A Quote
Quantity:
25 g
Request bulk or custom format
Chemical Identifiers
CAS4509-90-4
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to pale yellow
FormLiquid
Assay (GC)≥97.5%
Refractive Index1.4895-1.4935 @ 20?C
5-Bromovaleryl chloride was used in the preparation of aromatic azides and organic gelators.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
5-Bromovaleryl chloride was used in the preparation of aromatic azides and organic gelators.

Solubility
Reacts with water.

Notes
Moisture sensitive. Store away from water/ moisture, oxidizing agents, bases.
RUO – Research Use Only

General References:

  1. Rajavel Srinivasan; Mahesh Uttamchandani; Shao Q Yao. Rapid assembly and in situ screening of bidentate inhibitors of protein tyrosine phosphatases. Organic Letters. A. 2006, 8(4), 713-716.
  2. Myung Ho Hyun; Ok-Sang Jung, Hyun Jin Kim; Euh Duck Jeong. The role of the neutral and cationic gelators from (1S, 2S)-(-)-diphenylethylenediamine for the preparation of silica nano tubes.Bull. Korean Chem. Soc. 2009, 30 (7), 1641-1643.