Thermo Scientific Chemicals

4-Chloro-2-nitroaniline, 98%, Thermo Scientific Chemicals

Catalog number: A18143.0B
1000 g, Each
Thermo Scientific Chemicals

4-Chloro-2-nitroaniline, 98%, Thermo Scientific Chemicals

Catalog number: A18143.0B
1000 g, Each
Quantity
Catalog number: A18143.0B
also known as A18143-0B
Price (USD)
Quantity
-

Chemical Identifiers

CAS
89-63-4
Appearance (Color)
Orange
Assay (GC)
≥97.5%
Melting Point (clear melt)
112.0-118.0?C
Form
Powder
Identification (FTIR)
Conforms

Description

4-Chloro-2-nitroaniline is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Chloro-2-nitroaniline is used as pharmaceutical intermediate.

Solubility
Soluble in water (0.5 g/L).

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents and bases.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text