Dulcitol, 97%
Dulcitol, 97%
Dulcitol, 97%
Thermo Scientific Chemicals

Dulcitol, 97%

CAS: 608-66-2 | C6H14O6 | 182.172 g/mol
Quantity:
25 g
100 g
500 g
Catalog number A18402.36
also known as A18402-36
Price (JPY)
-
Quantity:
500 g
Request bulk or custom format
Chemical Identifiers
CAS608-66-2
IUPAC Namehexane-1,2,3,4,5,6-hexol
Molecular FormulaC6H14O6
InChI KeyFBPFZTCFMRRESA-UHFFFAOYNA-N
SMILESOCC(O)C(O)C(O)C(O)CO
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
FormCrystals or powder or crystalline powder
Assay (Silylated GC)≥96.0%
Melting Point (clear melt)185.0-194.0?C
Dulcitol is important in studies related to cataract formation, hepatosplenμly and mental retardation. It is used as a substrate to identify, differentiate and characterize galactitol dehydrogenase(s). It is also used as a reference compound in analytical procedures developed to analyze sugar alcohols in plants and food.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Dulcitol is important in studies related to cataract formation, hepatosplenµly and mental retardation. It is used as a substrate to identify, differentiate and characterize galactitol dehydrogenase(s). It is also used as a reference compound in analytical procedures developed to analyze sugar alcohols in plants and food.

Solubility
Soluble in water (30g/L at 14°C).

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents. Store under dry inert gas. It is hygroscopic in nature.
RUO – Research Use Only

General References:

  1. George S. Parks.; Hugh M. Huffman. Thermal Data on Organic Compounds. IV. The Heat Capacities, Entropies and Free Energies of Normal Propyl Alcohol, Ethyl ether and Dulcitol. J. Am. Chem. Soc. 1926, 48 (11), 2788-2793.
  2. Kikuo Takeda.; Shiro Ikushima.; Junichi Okuzaki.; Satoru Watanabe.; Taketoshi Fujimoto.; Taketoshi Nakahara. Inductively coupled plasma mass spectrometric determination of ultra-trace elements in electronic-grade water and chemicals using dulcitoly. Analytica Chimica Acta. 2001, 426 (1), 105-109.