Cyclohexylamine hydrobromide, 98%
Cyclohexylamine hydrobromide, 98%
Cyclohexylamine hydrobromide, 98%
Cyclohexylamine hydrobromide, 98%
Thermo Scientific Chemicals

Cyclohexylamine hydrobromide, 98%

CAS: 26227-54-3 | C6H14BrN | 180.089 g/mol
Quantity:
100 g
500 g
Catalog number A18439.22
also known as A18439-22
Price (JPY)
-
Request A Quote
Quantity:
100 g
Request bulk or custom format
Chemical Identifiers
CAS26227-54-3
IUPAC Namehydrogen cyclohexanamine bromide
Molecular FormulaC6H14BrN
InChI KeyQOHWJRRXQPGIQW-UHFFFAOYSA-N
SMILES[H+].[Br-].NC1CCCCC1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to pale cream
Assay (Titration ex Bromide)≥97.5 to ≤102.5%
FormCrystalline powder
Water Content (Karl Fischer Titration)<1.0%
Melting Point (clear melt)196-203?C
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only