2,4-Dinitrobenzenesulfonic acid sodium salt, 97%
2,4-Dinitrobenzenesulfonic acid sodium salt, 97%
2,4-Dinitrobenzenesulfonic acid sodium salt, 97%
Thermo Scientific Chemicals

2,4-Dinitrobenzenesulfonic acid sodium salt, 97%

CAS: 885-62-1 | C6H3N2NaO7S | 270.15 g/mol
Quantity:
5 g
25 g
Catalog number A18493.14
also known as A18493-14
Price (JPY)
-
Quantity:
25 g
Request bulk or custom format
Chemical Identifiers
CAS885-62-1
SpecificationsSpecification SheetSpecification Sheet
FormPowder
Sulfated Ash25.24 to 27.34% (theory 26.29%)
Appearance (Color)Yellow to orange
Assay (ex Sulphated Ash)≥ 96.0%
2,4-Dinitrobenzenesulfonic acid sodium salt is used in the synthesis of aromatic dimethylene spacer-inserted cyclodextrin derivatives.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,4-Dinitrobenzenesulfonic acid sodium salt is used in the synthesis of aromatic dimethylene spacer-inserted cyclodextrin derivatives.

Solubility
Soluble in water and methanol.

Notes
Hygroscopic. Moisture sensitive. Incompatible with strong oxidizing agents and strong bases.
RUO – Research Use Only

General References:

  1. Raza, R.; Saeed, A.; Lecka, J.; Sevigny, J.; Iqbal, J. Identification of Small Molecule Sulfonic Acids as Ecto-5'-Nucleotidase Inhibitors. Med. Chem. 2012, 8 (6), 1133-1139.
  2. Navid, F.; Boniotto, M.; Walker, C.; Ahrens, K.; Proksch, E.; Sparwasser, T.; Müller, W.; Schwarz, T.; Schwarz, A. Induction of Regulatory T Cells by a Murine beta-Defensin. J. Immunol. 2012, 188 (2), 735-743.