2-Fluoro-4-(trifluoromethyl)benzaldehyde, 97%
2-Fluoro-4-(trifluoromethyl)benzaldehyde, 97%
2-Fluoro-4-(trifluoromethyl)benzaldehyde, 97%
2-Fluoro-4-(trifluoromethyl)benzaldehyde, 97%
Thermo Scientific Chemicals

2-Fluoro-4-(trifluoromethyl)benzaldehyde, 97%

CAS: 89763-93-9 | C8H4F4O | 192.113 g/mol
Quantity:
1 g
5 g
25 g
Catalog number A18500.06
also known as A18500-06
Price (JPY)
-
Request A Quote
Quantity:
5 g
Request bulk or custom format
Chemical Identifiers
CAS89763-93-9
IUPAC Name2-fluoro-4-(trifluoromethyl)benzaldehyde
Molecular FormulaC8H4F4O
InChI KeyKFEHNXLFIGPWNB-UHFFFAOYSA-N
SMILESFC1=CC(=CC=C1C=O)C(F)(F)F
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
FormLiquid
Assay (GC)≥96.0%
Free acid (titration)≤3.0%
Refractive Index1.4475-1.4525 @ 20?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only