Dipropylene glycol, mixture of isomers, 99%
Dipropylene glycol, mixture of isomers, 99%
Dipropylene glycol, mixture of isomers, 99%
Thermo Scientific Chemicals

Dipropylene glycol, mixture of isomers, 99%

CAS: 25265-71-8 | C6H14O3 | 134.18 g/mol
Quantity:
500 g
2500 g
Catalog number A18716.0E
also known as A18716-0E
Price (JPY)
-
Quantity:
2500 g
Request bulk or custom format
Chemical Identifiers
CAS25265-71-8
IUPAC Name1-(1-hydroxypropoxy)propan-1-ol
Molecular FormulaC6H14O3
InChI KeyQYSXYAURTRCDJU-UHFFFAOYNA-N
SMILESCCC(O)OC(O)CC
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
FormLiquid
Assay (Silylated GC)≥98.5% (sum of isomers)
CommentProduct contains a mixture of three isomeric chemical compounds: 4-oxa-2,6-heptandiol, 2-(2-hydroxy-propoxy)-propan-1-ol and 2-(2-hydroxy-1-methyl-ethoxy)-propan-1-ol.
Identification (FTIR)Conforms
View more
Dipropylene glycol, mixture of isomers is used as a solvent, plasticizer, polymerization initiator and monomer. It acts as an additive used in perfumes, skin and hair care products. It is an active ingredient in commercial fog fluid, which is used as an entertainment in fog machines. Further, it is used as an intermediate in polyurethane polyols, alkyd resins and non-reinforced polyesters. In addition to this, it is used in dyes, paints, hydraulic brake fluids and cutting oils.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Dipropylene glycol, mixture of isomers is used as a solvent, plasticizer, polymerization initiator and monomer. It acts as an additive used in perfumes, skin and hair care products. It is an active ingredient in commercial fog fluid, which is used as an entertainment in fog machines. Further, it is used as an intermediate in polyurethane polyols, alkyd resins and non-reinforced polyesters. In addition to this, it is used in dyes, paints, hydraulic brake fluids and cutting oils.

Solubility
Miscible with cold water, hot water, methanol, diethyl ether and ethanol.

Notes
Hygroscopic. Incompatible with strong oxidizing agents.
RUO – Research Use Only

General References:

  1. Nicolae, M.; Oprea, F.; Fendu, E. M. Dipropylene glycol as a solvent for the extraction of aromatic hydrocarbons. Analysis and evaluation of the solvency properties and simulation of the extraction processes. Chem. Eng. Res. Des. 2015, 104, 287-295.
  2. Horbatenko, Y.; Pérez, J. P.; Hernández, P.; Swart, M.; Solà, M. Reaction Mechanisms for the Formation of Mono- And Dipropylene Glycol from the Propylene Oxide Hydrolysis over ZSM-5 Zeolite. J. Phys. Chem. C 2014, 118 (38), 21952-21962.