1,5-Diaminoanthraquinone, 90+%
1,5-Diaminoanthraquinone, 90+%
1,5-Diaminoanthraquinone, 90+%
Thermo Scientific Chemicals

1,5-Diaminoanthraquinone, 90+%

CAS: 129-44-2 | C14H10N2O2 | 238.246 g/mol
Quantity:
25 g
100 g
Catalog number A18840.14
also known as A18840-14
Price (JPY)
-
Quantity:
25 g
Request bulk or custom format
Chemical Identifiers
CAS129-44-2
IUPAC Name1,5-diamino-9,10-dihydroanthracene-9,10-dione
Molecular FormulaC14H10N2O2
InChI KeyVWBVCOPVKXNMMZ-UHFFFAOYSA-N
SMILESNC1=CC=CC2=C1C(=O)C1=CC=CC(N)=C1C2=O
View more
SpecificationsSpecification SheetSpecification Sheet
FormPowder
Identification (FTIR)Conforms
Appearance (Color)Dark red to red to dark brown to brown
Assay (HPLC)≥90.0%
1,5-Diaminoanthraquinone (DAAQ) may be used in the preparation of vertical nanowire arrays. It may be used in the synthesis of poly(1,5-diaminoanthraquinone) (PDAA) particles, via chemically oxidative polymerization of DAAQ.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,5-Diaminoanthraquinone (DAAQ) may be used in the preparation of vertical nanowire arrays. It may be used in the synthesis of poly(1,5-diaminoanthraquinone) (PDAA) particles, via chemically oxidative polymerization of DAAQ.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
RUO – Research Use Only

General References:

  1. Guohui Chang et. al. Synthesis of Pt nanoparticles decorated 1,5-diaminoanthraquinone nanofibers and their application toward catalytic reduction of 4-nitrophenol. Journal of Nanoscience and Nanotechnology, 2012, 12 (9), 7075-7080.
  2. Xin-Gui Li; Hu Li; Mei-Rong Huang. Productive synthesis and properties of polydiaminoanthraquinone and its pure self-stabilized nanoparticles with widely adjustable electroconductivity. Chemistry: A European Journal, 2007, 13(31), 8884-8896.