8-Amino-2-naphthol, 98%
8-Amino-2-naphthol, 98%
8-Amino-2-naphthol, 98%
Thermo Scientific Chemicals

8-Amino-2-naphthol, 98%

CAS: 118-46-7 | C10H9NO | 159.19 g/mol
Quantity:
1 g
5 g
25 g
Catalog number A19404.14
also known as A19404-14
Price (JPY)
-
Quantity:
25 g
Request bulk or custom format
Chemical Identifiers
CAS118-46-7
IUPAC Name8-aminonaphthalen-2-ol
Molecular FormulaC10H9NO
InChI KeyKVHHMYZBFBSVDI-UHFFFAOYSA-N
SMILESNC1=C2C=C(O)C=CC2=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
FormPowder
Identification (FTIR)Conforms
Appearance (Color)Grey to brown to dark brown
Assay (HPLC)≥96.0%
Melting Point (clear melt)197-207°C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only