1,5-Dichloroanthraquinone, 96%
1,5-Dichloroanthraquinone, 96%
1,5-Dichloroanthraquinone, 96%
Thermo Scientific Chemicals

1,5-Dichloroanthraquinone, 96%

CAS: 82-46-2 | C14H6Cl2O2 | 277.1 g/mol
Quantity:
50 g
250 g
Catalog number A19534.18
also known as A19534-18
Price (JPY)
-
Quantity:
50 g
Request bulk or custom format
Chemical Identifiers
CAS82-46-2
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Yellow
FormPowder
Assay (HPLC)≥95.0%
Identification (FTIR)Conforms
Melting Point (clear melt)241.0-251.0?C
1,5-Dichloroanthraquinone undergoes a solid-state transformation at 470 K. The two modifications are very similar, except that a is greater than b above the transition temperature and the inverse is at room temperature.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,5-Dichloroanthraquinone undergoes a solid-state transformation at 470 K. The two modifications are very similar, except that a is greater than b above the transition temperature and the inverse is at room temperature.

Solubility
Insoluble in water.

Notes
Keep container tightly closed. Store away from oxidizing agents.
RUO – Research Use Only

General References:

  1. P.-W. Wang. A solid-state transformation in 1,5-dichloroanthraquinone. J. Appl. Cryst. 1979, 12, 239
  2. Edward H. Ruediger.; Magdy L. Kaldas.; Sham S. Gandhi.; Cristi Fedryna.; Martin S. Gibson. Reactions of 1,5-dichloroanthraquinone with nucleophiles. J. Org. Chem. 1980, 45, (10), 1974-1978