1-Tetradecanol, 97+%
1-Tetradecanol, 97+%
1-Tetradecanol, 97+%
Thermo Scientific Chemicals

1-Tetradecanol, 97+%

CAS: 112-72-1 | C14H30O | 214.39 g/mol
Quantity:
100 g
500 g
2500 g
Catalog number A19638.0E
also known as A19638-0E
Price (JPY)
-
Quantity:
2500 g
Request bulk or custom format
Chemical Identifiers
CAS112-72-1
IUPAC Nametetradecan-1-ol
Molecular FormulaC14H30O
InChI KeyHLZKNKRTKFSKGZ-UHFFFAOYSA-N
SMILESCCCCCCCCCCCCCCO
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
FormGranules, prills or crystalline powder
Assay (GC)>97.0%
1-Tetradecanol is used as an ingredient in cosmetics such as cold creams. It is an active intermediate in the chemical synthesis of sulfated alcohol. It is also employed in the fabrication of temperature-regulated drug release system based on phase-change materials. It plays a vital role in filling the hollow interiors of gold nanocages in the fabrication of new theranostic system, which has unique feature of photoacoustic imaging.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Tetradecanol is used as an ingredient in cosmetics such as cold creams. It is an active intermediate in the chemical synthesis of sulfated alcohol. It is also employed in the fabrication of temperature-regulated drug release system based on phase-change materials. It plays a vital role in filling the hollow interiors of gold nanocages in the fabrication of new theranostic system, which has unique feature of photoacoustic imaging.

Solubility
Insoluble in water. Soluble in diethyl ether. Slightly soluble in ethanol.

Notes
Incompatible with strong oxidizing agents, acid chlorides and acid anhydrides. Store in a cool place.
RUO – Research Use Only

General References:

  1. Tian, T.; Song, J.; Niu, L.; Feng, R. Preparation and properties of 1-tetradecanol/1,3:2,4-di-(3,4-dimethyl) benzylidene sorbitol gelatinous form-stable phase change materials. Thermochim. Acta 2013, 554, 54-58.
  2. Corberan, V. C.; Gomez-Aviles, A.; Martinez-Gonzalez, S.; Ivanova, S.; Dominguez, M. I.; Gonzalez-Perez, M. E. Heterogeneous selective oxidation of fatty alcohols: Oxidation of 1-tetradecanol as a modelsubstrate. Catal. Today 2014, 238, 49-53.