1,2-Bis(methanesulfonamido)benzene, 97%
1,2-Bis(methanesulfonamido)benzene, 97%
1,2-Bis(methanesulfonamido)benzene, 97%
Thermo Scientific Chemicals

1,2-Bis(methanesulfonamido)benzene, 97%

CAS: 7596-80-7 | C8H12N2O4S2 | 264.31 g/mol
Quantity:
2.5 g
10 g
Catalog number A19830.09
also known as A19830-09
Price (JPY)
-
Quantity:
10 g
Request bulk or custom format
Chemical Identifiers
CAS7596-80-7
SpecificationsSpecification SheetSpecification Sheet
FormPowder
Melting Point (clear melt)209.5-218.5?C
Assay (Aqueous acid-base Titration)≥96.0 to ≤104.0%
Appearance (Color)Pink
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only