4-Fluorophenylsulfonylacetonitrile, 97%
4-Fluorophenylsulfonylacetonitrile, 97%
4-Fluorophenylsulfonylacetonitrile, 97%
4-Fluorophenylsulfonylacetonitrile, 97%
Thermo Scientific Chemicals

4-Fluorophenylsulfonylacetonitrile, 97%

CAS: 32083-66-2 | C8H6FNO2S | 199.199 g/mol
Quantity:
1 g
5 g
Catalog number A19916.03
also known as A19916-03
Price (JPY)
-
Request A Quote
Quantity:
1 g
Request bulk or custom format
Chemical Identifiers
CAS32083-66-2
IUPAC Name2-(4-fluorobenzenesulfonyl)acetonitrile
Molecular FormulaC8H6FNO2S
InChI KeyWBXJZTLPEMITJL-UHFFFAOYSA-N
SMILESFC1=CC=C(C=C1)S(=O)(=O)CC#N
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)≥96.0%
Melting Point (clear melt)87.0-93.0?C
FormCrystals or powder or crystalline powder or flakes
Appearance (Color)White
4-Fluorophenylsulfonylacetonitrile is used to produce 3-(4-fluoro-benzenesulfonyl)-thiophen-2-ylamine with [1,4]dithiane-2,5-diol. It is also used as pharmaceutical intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Fluorophenylsulfonylacetonitrile is used to produce 3-(4-fluoro-benzenesulfonyl)-thiophen-2-ylamine with [1,4]dithiane-2,5-diol. It is also used as pharmaceutical intermediates.

Notes
Incompatible with oxidizing agents. Store in a cool, dry conditions in a well sealed container.
RUO – Research Use Only