ABY™ Dye Spectral Calibration Plate for Multiplex qPCR, 96-well - FAQs
ABY™ Dye Spectral Calibration Plate for Multiplex qPCR, 96-well - FAQs
View additional product information for ABY™ Dye Spectral Calibration Plate for Multiplex qPCR, 96-well - FAQs (A24738)
1 product FAQs found
What are the excitation and emission profiles for ABY, JUN, Cyanine 5, and Cyanine 5.5 dyes?
ABY dye has an excitation wavelength of 568 nm and emission wavelength of 583 nm.
JUN dye has an excitation wavelength of 606 nm and emission wavelength of 618 nm.
Cynanine 5 dye has an excitation wavelength of 649 nm and emission wavelength of 667 nm.
Cyanine 5.5 dye has an excitation wavelength of 678 nm and emission wavelength of 695 nm.
Please refer to your instrument user guide for additional information on compatibility with different dye channels.
Find additional tips, troubleshooting help, and resources within our Real-Time PCR and Digital PCR Instruments Support Center.