Alexa Fluor™ 647-aha-dUTP, 1 mM in TE buffer
Invitrogen™

Alexa Fluor™ 647-aha-dUTP, 1 mM in TE buffer

Labeled 5-aminohexylacrylamido-dUTP (aha-dUTP) nucleotides are modified at the C-5 position of uridine with a unique hexylacrylamide linker, which serves asRead more
Have Questions?
Catalog NumberQuantity
A3276350 μL
Catalog number A32763
Price (USD)
912.00
Each
Quantity:
50 μL
Price (USD)
912.00
Each
Labeled 5-aminohexylacrylamido-dUTP (aha-dUTP) nucleotides are modified at the C-5 position of uridine with a unique hexylacrylamide linker, which serves as a spacer between the nucleotide and the dye. This spacer reduces interactions between the nucleotide and the dye, resulting in brighter conjugates and increased hapten accessibility for secondary detection reagents. Labeled nucleotides can be used to generate labeled nucleic acid hybridization probes for many molecular biology and molecular cytogenetics applications, including multicolor techniques.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Concentration1 mM
For Use With (Application)Molecular Biology
Label or DyeAlexa Fluor Dye
ModificationAHA (5-aminohexylacrylamido)
Product LineAlexa Fluor
Product TypeLabeled Nucleotide
Quantity50 μL
Shipping ConditionWet Ice
Unit SizeEach
Contents & Storage
Store in freezer (-5 to -30°C) and protect from light.

Frequently asked questions (FAQs)

Can dU-containing DNA produced by amplification with dUTP be cut by restriction enzymes?

You will need to confirm with the manufacturer of the restriction enzyme you are using to find out whether or not it can recognize dUTP-containing DNA.

Find additional tips, troubleshooting help, and resources within our PCR and cDNA Synthesis Support Center.