Ion 510™ Chip Kit - FAQs
Ion 510™ Chip Kit - FAQs
View additional product information for Ion 510™ Chip Kit - FAQs (A34292)
25 product FAQs found
What is the maximum throughput/day for the Ion S5 GeneStudio Prime System?
The Ion S5 GeneStudio Prime System has a maximum throughput/day of 50 Gb (2 Ion 550 Chips).
Find additional tips, troubleshooting help, and resources within our Next-Generation Sequencing Support Center.
How long does it take to run an Ion 550 Chip on the Ion S5 GeneStudio Prime System?
The Ion S5 GeneStudio Prime System takes 8.5 hours to run an Ion 550 Chip.
Find additional tips, troubleshooting help, and resources within our Next-Generation Sequencing Support Center.
How long does it take to run an Ion 540 Chip on the Ion S5 GeneStudio Prime System?
The Ion S5 GeneStudio Prime System takes 6.5 hours to run an Ion 540 Chip.
Find additional tips, troubleshooting help, and resources within our Next-Generation Sequencing Support Center.
What chips are compatible with the Ion GeneStudio S5 Prime System?
Ion 510 Chip, Ion 520 Chip, Ion 530 Chip, Ion 540 Chip, and Ion 550 Chip are compatible with the Ion GeneStudio S5 Prime System.
Find additional tips, troubleshooting help, and resources within our Next-Generation Sequencing Support Center.
Is the Ion GeneStudio S5 Prime System compatible with the Ion One Touch 2 System?
Yes, the Ion GeneStudio S5 Prime System compatible with the Ion One Touch 2 System.
Find additional tips, troubleshooting help, and resources within our Next-Generation Sequencing Support Center.
Is the Ion GeneStudio S5 Prime System compatible with the Ion Chef System?
Yes, the Ion GeneStudio S5 Prime System is compatible with the Ion Chef System.
Find additional tips, troubleshooting help, and resources within our Next-Generation Sequencing Support Center.
Does the Ion GeneStudio S5 Prime System have a separate server?
Yes, the Ion GeneStudio S5 Prime System has a separate server.
Find additional tips, troubleshooting help, and resources within our Next-Generation Sequencing Support Center.
What is the server storage size on the Ion GeneStudio S5 Prime System?
The server storage size is 25 TB.
Find additional tips, troubleshooting help, and resources within our Next-Generation Sequencing Support Center.
What is the maximum throughput/day for the Ion GeneStudio S5 Plus System?
The Ion GeneStudio S5 Plus System has a maximum throughput/day of 30 Gb (2 Ion 540 Chips).
Find additional tips, troubleshooting help, and resources within our Next-Generation Sequencing Support Center.
How long does it take to run an Ion 550 Chip on the Ion GeneStudio S5 Plus System?
The Ion GeneStudio S5 Plus System takes 11.5 hours to run an Ion 550 Chip.
Find additional tips, troubleshooting help, and resources within our Next-Generation Sequencing Support Center.
How long does it take to run an Ion 540 Chip on the Ion GeneStudio S5 Plus System?
The Ion GeneStudio S5 Plus System takes 10 hours to run an Ion 540 Chip.
Find additional tips, troubleshooting help, and resources within our Next-Generation Sequencing Support Center.
What chips are compatible with the Ion GeneStudio S5 Plus System?
Ion 510 Chip, Ion 520 Chip, Ion 530 Chip, Ion 540 Chip, and Ion 550 Chip are compatible with the Ion GeneStudio S5 Plus System.
Find additional tips, troubleshooting help, and resources within our Next-Generation Sequencing Support Center.
Is the Ion GeneStudio S5 Plus System compatible with the Ion One Touch 2 System?
Yes, the Ion GeneStudio S5 Plus System compatible with the Ion One Touch 2 System.
Find additional tips, troubleshooting help, and resources within our Next-Generation Sequencing Support Center.
Is the Ion GeneStudio S5 Plus System compatible with the Ion Chef System?
Yes, the Ion GeneStudio S5 Plus System compatible with the Ion Chef System.
Find additional tips, troubleshooting help, and resources within our Next-Generation Sequencing Support Center.
Does the Ion GeneStudio S5 Plus System have a separate server?
No, the server is contained inside the sequencer itself.
Find additional tips, troubleshooting help, and resources within our Next-Generation Sequencing Support Center.
What is the server storage size on the Ion GeneStudio S5 Plus System?
The server storage size on the Ion GeneStudio S5 Plus System is 24 TB.
Find additional tips, troubleshooting help, and resources within our Next-Generation Sequencing Support Center.
Is the Ion GeneStudio S5 System compatible with the Ion One Touch 2 System?
Yes, the Ion GeneStudio S5 System compatible with the Ion One Touch 2 System.
Find additional tips, troubleshooting help, and resources within our Next-Generation Sequencing Support Center.
Is the Ion GeneStudio S5 System compatible with the Ion Chef System?
Yes, the Ion GeneStudio S5 System compatible with the Ion Chef System.
Find additional tips, troubleshooting help, and resources within our Next-Generation Sequencing Support Center.
What is the server storage size on the Ion GeneStudio S5 System?
The server storage size is 12 TB.
Find additional tips, troubleshooting help, and resources within our Next-Generation Sequencing Support Center.
How long does analysis take on the Ion GeneStudio S5 System?
Please see the following table (https://www.thermofisher.com/us/en/home/life-science/sequencing/next-generation-sequencing/ion-torrent-next-generation-sequencing-workflow/ion-torrent-next-generation-sequencing-run-sequence/ion-s5-ngs-targeted-sequencing/ion-s5-specifications.html).
Find additional tips, troubleshooting help, and resources within our Next-Generation Sequencing Support Center.
Can the Ion GeneStudio S5 System run two chips back-to-back?
No, the analysis of the first chip must complete before the sequencing run of the second chip begins.
Find additional tips, troubleshooting help, and resources within our Next-Generation Sequencing Support Center.
Does the Ion GeneStudio S5 System have a separate analysis server?
No, the Ion GeneStudio S5 System's analysis server is contained within the sequencer itself.
Find additional tips, troubleshooting help, and resources within our Next-Generation Sequencing Support Center.
How long does it take to run an Ion 540 Chip on the Ion GeneStudio S5 System?
The Ion GeneStudio S5 System takes 19 hours to run and analyze an Ion 540 Chip.
Find additional tips, troubleshooting help, and resources within our Next-Generation Sequencing Support Center.
What is the maximum throughput/day for the Ion GeneStudio S5 System?
The Ion GeneStudio S5 System has a maximum throughput/day of 15 Gb.
Find additional tips, troubleshooting help, and resources within our Next-Generation Sequencing Support Center.
What chips are compatible with the Ion GeneStudio S5 System?
Ion 510 Chip, Ion 520 Chip, Ion 530 Chip, and Ion 540 Chip are compatible with the Ion GeneStudio S5 System.
Find additional tips, troubleshooting help, and resources within our Next-Generation Sequencing Support Center.