Ion 510™ Chip Kit
Ion Torrent™
Ion 510™ Chip Kit
The Ion 510 Chip Kit contains 8 barcoded chips for sample tracking and sequencing with the Ion S5 and Ion S5 XL sequencing systems.
Have Questions?
Catalog NumberQuantity
A342928 Chips
Catalog number A34292
Price (USD)
3,740.00
Each
Quantity:
8 Chips
Price (USD)
3,740.00
Each
The Ion 510 Chip Kit contains 8 barcoded chips for sample tracking and sequencing with the Ion S5 and Ion S5 XL sequencing systems. The Ion 510 Chip electronically detects polymerase-driven base incorporation without the use of fluorescence. By eliminating the use of an optical detection system, this advance in next-generation sequencing technology allows for rapid sequencing times in as little as 2.5 hours for 200 bp sequencing and 4 hours for 400 bp sequencing.
The Ion 510 Chip Kit offers:
  • Rapid run times of only 2.5–4 hours, generating 2–3 million reads
  • Read lengths up to 400 bp to support a variety of research applications, including targeted gene sequencing and microbial sequencing
  • Easy handling and loading of templated products for sequencing
  • Compatibility with current library preparation methods
Notes:
  • The Ion 510 Chip is only compatible with the Ion S5 and Ion S5 XL systems.
  • The Ion 510 Chip is only compatible with the Ion 510 & Ion 520 & Ion 530 Kit-Chef (Cat. Nos. A34019 and A34461).
  • The Ion 510 Chip is not validated for the Ion OneTouch 2 workflow.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Number of Reads per Chip2-3M
Optimal Reaction Temperature20°C to 25°C
Product LineIon Chef, Ion Torrent
Quantity8 Chips
Shipping ConditionRoom Temperature
Unit SizeEach
Contents & Storage
Store at room temperature.
Frequently asked questions (FAQs)
What is the maximum throughput/day for the Ion S5 GeneStudio Prime System?
The Ion S5 GeneStudio Prime System has a maximum throughput/day of 50 Gb (2 Ion 550 Chips).
Find additional tips, troubleshooting help, and resources within our Next-Generation Sequencing Support Center.
How long does it take to run an Ion 550 Chip on the Ion S5 GeneStudio Prime System?
The Ion S5 GeneStudio Prime System takes 8.5 hours to run an Ion 550 Chip.
Find additional tips, troubleshooting help, and resources within our Next-Generation Sequencing Support Center.
How long does it take to run an Ion 540 Chip on the Ion S5 GeneStudio Prime System?
The Ion S5 GeneStudio Prime System takes 6.5 hours to run an Ion 540 Chip.
Find additional tips, troubleshooting help, and resources within our Next-Generation Sequencing Support Center.
What chips are compatible with the Ion GeneStudio S5 Prime System?
Ion 510 Chip, Ion 520 Chip, Ion 530 Chip, Ion 540 Chip, and Ion 550 Chip are compatible with the Ion GeneStudio S5 Prime System.
Find additional tips, troubleshooting help, and resources within our Next-Generation Sequencing Support Center.
Is the Ion GeneStudio S5 Prime System compatible with the Ion One Touch 2 System?
Yes, the Ion GeneStudio S5 Prime System compatible with the Ion One Touch 2 System.
Find additional tips, troubleshooting help, and resources within our Next-Generation Sequencing Support Center.