Product Image
Applied Biosystems™
Variant Reporter™ v3 Upgrade from v2
This is the v2 to v3 upgrade for Variant Reporter Software.
Have Questions?
Catalog NumberQuantity
A411151 CD
Catalog number A41115
Price (USD)
-
Quantity:
1 CD
This is the v2 to v3 upgrade for Variant Reporter Software. This software performs comparative sequencing, also known as direct sequencing, medical sequencing, PCR sequencing, and resequencing with DNA sequencing files. The software is designed for reference-based and non-reference-based analysis such as mutation detection and analysis, SNP discovery and validation, and sequence confirmation. The robust algorithms will call SNPs, mutations, insertions, deletions, and heterozygous insertions/deletions for data generated using the Applied Biosystems genetic analyzers.
Note:
This product can only be used to upgrade Variant Reporter Software v2 installed on standalone computers (computers separate from the instrument data collection system). Please contact your Sales Representative to upgrade the data collection system and secondary analysis software running on the data collection computer.
Specifications
Data EntrySequencing traces
Data OutputsVariant Reports with sequence comparison metrics
For Use With (Equipment)310 Genetic Analyzer, 3100 Genetic Analyzer, 3100-Avant Genetic Analyzer, 3130xl Genetic Analyzer, 3730xl DNA Analyzer, 3500 Genetic Analyzer, 3730 DNA Analyzer
LicenseStatic
Operating SystemWindows 10 Pro/IoT, Windows 7
Product TypeVariant Reporter V3 Upgrade From V2
Quantity1 CD
Software CategoryUpgrade
System RequirementsComputer: Core 2 Duo E8400/3 GHz; 2 GB RAM
Product LineVariant Reporter
Software TypeComparative Sequencing
Unit SizeEach
Contents & Storage
Contains one software CD.
Frequently asked questions (FAQs)
Which instruments does Variant Reporter Software v3.0 support?
Variant Reporter Software v3.0 fully supports data generated from the following instruments running all versions of Data Collection Software:
- Applied Biosystems SeqStudio Genetic Analyzer
- Applied Biosystems 3500 Genetic Analyzer
- Applied Biosystems 3500xL Genetic Analyzer
- Applied Biosystems 3130 Genetic Analyzer
- Applied Biosystems 3130xl Genetic Analyzer
- Applied Biosystems 3730 DNA Analyzer
- Applied Biosystems 3730xl DNA Analyzer
Find additional tips, troubleshooting help, and resources within our Capillary Electrophoresis Applications Support Center.
What has been updated in Variant Reporter Software v3.0?
The primary updates in Variant Reporter Software v3.0 are the following:
- Variant Reporter Software v3.0 supports installation on Microsoft Windows 7 (64-bit and 32-bit) and Microsoft Windows 10 Professional (64-bit) operating systems.
- Variant Reporter Software v3.0 uses KB Basecaller version 1.4.2.4 for basecalling.
Find additional tips, troubleshooting help, and resources within our Capillary Electrophoresis Applications Support Center.
How can I import my data from Variant Reporter Software?
For importing data, there are many ways to do so:
More information on backing up the Data Store can be found in the Variant Reporter Help Contents under the keyword “Back-up”.
1. Select the Data Store where you would like the data imported to. To choose a Data Store:
a. Click the Manage button in the Data Store section of the Dashboard View.
b. Highlight the Data Store you wish to use and click on Change Location to select it. Click Close to close the window.
Find additional tips, troubleshooting help, and resources within our Capillary Electrophoresis Software Support Center.
How can I archive a selected project in the Variant Reporter Software?
To archive a Variant Reporter software project from the Dashboard:
1. Open the Dashboard View.
2. In the Project tab select the project you want to archive, then click .
3. In the Archive Destination dialog box, browse to the folder where you want the project archived.
4. Click OK. The project file you archived will no longer appear in the Variant Reporter Dashboard.
More information on archiving Project data can be found in the Variant Reporter Help Contents under the keyword “Archive”.
Find additional tips, troubleshooting help, and resources within our Capillary Electrophoresis Software Support Center.
How can I back up the Data Store in the Variant Reporter Software?
To back up the Data Store:
1. Open the Dashboard View.
2. Click .
3. In the Browse for Back-up Folder Destination dialog box, select the folder that you want to back up the Data Store to, then click OK. Your project will start to get backed up.
Note: All associated Projects, Analysis Parameters, and References contained in the current Data Store will be copied to the folder you selected when you back up the Variant Reporter Data Store.
Find additional tips, troubleshooting help, and resources within our Capillary Electrophoresis Software Support Center.