TrueMark™ MSI Assay
TrueMark™ MSI Assay
TrueMark™ MSI Assay
Applied Biosystems™

TrueMark™ MSI Assay

The Applied Biosystems TrueMark MSI Assay is a low-input, single-amplification solution for the rapid identification of microsatellite instability (MSI) inRead more
Have Questions?
Catalog NumberNo. of Reactions
A45295100
Catalog number A45295
Price (USD)
3,580.00
Each
No. of Reactions:
100
Price (USD)
3,580.00
Each
The Applied Biosystems TrueMark MSI Assay is a low-input, single-amplification solution for the rapid identification of microsatellite instability (MSI) in formalin-fixed paraffin-embedded (FFPE) samples from multiple tumor tissue types for research applications. The TrueMark MSI Assay leverages an expanded 6-dye set to incorporate additional markers. The TrueMark MSI Assay is best suited for the analysis of colon cancer samples for research. After careful verification with your samples and your markers of interest, the free auto-calling software may be used without a tumor-normal match for colon cancer samples. The assay is compatible with the Applied Biosystems SeqStudio, SeqStudio Flex, and 3500 Series genetic analyzers.

MSI analysis software
The associated TrueMark MSI Analysis Software (included at no additional charge) is designed specifically for the TrueMark MSI Assay to enable automated calling of markers and samples, offering greater ease of analysis by reducing the manual QC, analysis, and review required to determine sample status. The automated calling is best used after a verification study and can be used with tumor-normal matched samples as well as without tumor-normal pairs.

Download TrueMark MSI Analysis Software ›

The TrueMark MSI Assay features include:
• Expanded coverage—13 MSI markers
    --BAT-25, BAT-26, BAT-40, CAT-25, NR-21, NR-22, NR-24, NR-27 (publicly known MSI markers)
    --ABI-16, ABI-17, ABI-19, ABI-20A, ABI-20B (MSI markers proprietary to Thermo Fisher Scientific)
    --PentaD, TH-01 (sample ID markers)
• Optional normal control—no requirement for use of tumor-normal match for interpretation
• Low input—use as little as 2 ng FFPE-derived DNA
• Automated calling—analysis software includes automated calling of markers and samples to accelerate initial data interpretation
• Simple analysis software—streamline QC, analysis, and review of samples

Analysis software for download
• TrueMark MSI Analysis Software

Supplied reagents
• TrueMark MSI Primer mix
• TrueMark MSI Master mix
• TrueMark MSI No Template Control
• TrueMark MSI Amplification Control

Required reagents not supplied in the TrueMark MSI Assay
• GeneScan 600 LIZ Size Standard v2.0
• Hi-Di Formamide
• DS-36 Matrix Standard (Dye Set J6)

The TrueMark MSI Assay has been tested with RecoverAll Total Nucleic Acid Isolation Kit for FFPE and Qiagen QIAamp™ DSP DNA FFPE Tissue Kit for sample preparation. Please note these are not included in the kit.

For Research Use Only. Not for use in diagnostic procedures.
Specifications
For Use With (Application)Sequencing
No. of Reactions100
Product LineTrueMark
Product TypeMSI Assay
Shipping ConditionDry Ice
Test TypeOncology
Unit SizeEach
Contents & Storage
• TrueMark MSI Assay Master Mix
• TrueMark MSI Assay Primer Mix
• TrueMark MSI Assay Amplification Control
• TrueMark MSI Assay No-Template Control

Store at -15 to -25°C, then 2–8°C after first use.

Frequently asked questions (FAQs)

Can I freeze TrueMark MSI Assay (Cat. No. A45295) after my initial use?

We recommend storing TrueMark MSI Assay in the refrigerator after initial use because there are a lot of dye-labeled primers in the mix. Freeze-thaws could cause the dye to break down.

Find additional tips, troubleshooting help, and resources within our Capillary Electrophoresis Applications Support Center.

Can I use GeneMapper Software to analyze data generated with the TrueMark MSI Assay?

TrueMark MSI Assay data can be analyzed in GeneMapper Software, but a tumor-normal match must be run and there is no automated calling. We recommend using the full solution for the TrueMark MSI Assay which includes the new TrueMark MSI Assay Analysis Software.

Find additional tips, troubleshooting help, and resources within our Capillary Electrophoresis Applications Support Center.

Can I use the TrueMark MSI Assay Analysis Software with any other MSI kits from other known vendors or with an MSI assay that I develop myself?

No, the TrueMark MSI Assay Analysis Software is designed exclusively for use with and custom designed around the panel used in our TrueMark MSI Assay.

Find additional tips, troubleshooting help, and resources within our Capillary Electrophoresis Applications Support Center.

Where can I find the TrueMark MSI Assay Quick Reference Guide?

You may find the TrueMark MSI Assay Quick Reference Guide here: https://assets.thermofisher.com/TFS-Assets/LSG/manuals/MAN0018869_MSI%20Assay_QR.pdf

Find additional tips, troubleshooting help, and resources within our Capillary Electrophoresis Applications Support Center.

Where can I find the TrueMark MSI Assay User Guide?

You may find the TrueMark MSI Assay User Guide here: https://assets.thermofisher.com/TFS-Assets/LSG/manuals/MAN0018868_MSI%20Assay_UG.pdf

Find additional tips, troubleshooting help, and resources within our Capillary Electrophoresis Applications Support Center.