Barium Chloride Dihydrate, Certified AR for Analysis, Fisher Chemical™
Barium Chloride Dihydrate, Certified AR for Analysis, Fisher Chemical™
Barium Chloride Dihydrate, Certified AR for Analysis, Fisher Chemical™
Fisher Chemical™

Barium Chloride Dihydrate, Certified AR for Analysis, Fisher Chemical™

Catalog NumberQuantity
B-0550-53
also known as B/0550/53
500 g
Catalog number B-0550-53
also known as B/0550/53
Price (JPY)
-
Quantity:
500 g
Request bulk or custom format
Specifications
Boiling Point1560°C
CAS10326-27-9
ColorWhite
Formula Weight244.28g/mol
IUPAC Namebarium(2+) dihydrate dichloride
View more
Barium Chloride Dihydrate, Certified AR for Analysis