Boric Acid, Certified AR for Analysis, Fisher Chemical™
Boric Acid, Certified AR for Analysis, Fisher Chemical™
Boric Acid, Certified AR for Analysis, Fisher Chemical™
Fisher Chemical™

Boric Acid, Certified AR for Analysis, Fisher Chemical™

H3BO3, CAS Number-10043-35-3, 500g, White, 61.83g/mol, 169 deg.C, 11337, Plastic powder jar, 3.8 to 4.8, Powder Solid, 2.7mbar at 20 deg.C | CAS: 10043-35-3 | BH3O3 | 61.83 g/mol
Quantity:
500 g
1 kg
Catalog number B-3800-60
also known as B/3800/60
Price (JPY)
-
Quantity:
1 kg
Request bulk or custom format
Specifications
CAS10043-35-3
ColorWhite
Formula Weight61.83g/mol
IUPAC Nameboric acid
InChI KeyKGBXLFKZBHKPEV-UHFFFAOYSA-N
View more
H3BO3, CAS Number-10043-35-3, 500g, White, 61.83g/mol, 169 deg.C, 11337, Plastic powder jar, 3.8 to 4.8, Powder Solid, 2.7mbar at 20 deg.C