2,6-Dimethylanisole, 98+%
2,6-Dimethylanisole, 98+%
2,6-Dimethylanisole, 98+%
2,6-Dimethylanisole, 98+%
Thermo Scientific Chemicals

2,6-Dimethylanisole, 98+%

CAS: 1004-66-6 | C9H12O | 136.19 g/mol
Quantity:
25 g
100 g
Catalog number B20365.14
also known as B20365-14
Price (JPY)
-
Request A Quote
Quantity:
25 g
Request bulk or custom format
Chemical Identifiers
CAS1004-66-6
IUPAC Name2-methoxy-1,3-dimethylbenzene
Molecular FormulaC9H12O
InChI KeyGFNZJAUVJCGWLW-UHFFFAOYSA-N
SMILESCOC1=C(C)C=CC=C1C
View more
SpecificationsSpecification SheetSpecification Sheet
FormLiquid
Refractive Index1.5010-1.5050 @ 20?C
Appearance (Color)Clear colorless to yellow
Assay (GC)≥98.0%
2,6-Dimethylanisole is used as a pharmaceutical intermediate. Methoxymetacyclophanes is prepared from 2,6-dimethylanisole.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,6-Dimethylanisole is used as a pharmaceutical intermediate. Methoxymetacyclophanes is prepared from 2,6-dimethylanisole.

Solubility
Insoluble in water.

Notes
Store in cool, dry place in a well sealed container. Store away from oxidizing agents.
RUO – Research Use Only

General References:

  1. Leslie R. Dix and Roy B. Moodie. Nitrosation and nitrous acid-catalysed nitration of anisole and 2,6-dimethylanisole. J. Chem. Soc., Perkin Trans. 2. 1986, (7), 1097-1101.
  2. Robert B. Bates; Sanjeev Gangwar; Vinayak V. Kane; Kessara Suvannachut; Stuart R. Taylor. Methoxymetacyclophanes from 2,6-dimethylanisole. J. Org. Chem. 1982, 56 (5), 1696-1699.