2-Hydrazino-2-imidazoline hydrobromide, 98%
2-Hydrazino-2-imidazoline hydrobromide, 98%
2-Hydrazino-2-imidazoline hydrobromide, 98%
Thermo Scientific Chemicals

2-Hydrazino-2-imidazoline hydrobromide, 98%

CAS: 55959-84-7 | C3H9BrN4 | 181.037 g/mol
Quantity:
2.5 g
10 g
Catalog number B20386.88
also known as B20386-88
Price (JPY)
-
Quantity:
2.5 g
Request bulk or custom format
Chemical Identifiers
CAS55959-84-7
IUPAC Namehydrogen 2-hydrazinyl-4,5-dihydro-1H-imidazole bromide
Molecular FormulaC3H9BrN4
InChI KeyHIJRJVVSABWXKG-UHFFFAOYSA-N
SMILES[H+].[Br-].NNC1=NCCN1
View more
SpecificationsSpecification SheetSpecification Sheet
FormCrystals or powder or crystalline powder
Identification (FTIR)Conforms
Assay (Titration ex Bromide)≥97.5 to ≤102.5%
Appearance (Color)White to cream or pale grey
Melting Point (clear melt)180.0-186.0?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only