8-Chloro-2-methylquinoline, 98%
8-Chloro-2-methylquinoline, 98%
8-Chloro-2-methylquinoline, 98%
8-Chloro-2-methylquinoline, 98%
Thermo Scientific Chemicals

8-Chloro-2-methylquinoline, 98%

CAS: 3033-82-7 | C10H8ClN | 177.631 g/mol
Quantity:
1 g
5 g
Catalog number B20504.06
also known as B20504-06
Price (JPY)
-
Request A Quote
Quantity:
5 g
Request bulk or custom format
Chemical Identifiers
CAS3033-82-7
IUPAC Name8-chloro-2-methylquinoline
Molecular FormulaC10H8ClN
InChI KeyVVLYDFPOGMTMFJ-UHFFFAOYSA-N
SMILESCC1=CC=C2C=CC=C(Cl)C2=N1
View more
SpecificationsSpecification SheetSpecification Sheet
FormCrystals or powder or crystalline powder or fused solid
Assay (GC)≥97.5%
Appearance (Color)Pale cream to cream
Melting Point (clear melt)64.0-70.0?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only