1,5-Dibromo-3-methylpentane, 98+%
1,5-Dibromo-3-methylpentane, 98+%
1,5-Dibromo-3-methylpentane, 98+%
1,5-Dibromo-3-methylpentane, 98+%
Thermo Scientific Chemicals

1,5-Dibromo-3-methylpentane, 98+%

CAS: 4457-72-1 | C6H12Br2 | 243.97 g/mol
Quantity:
5 g
25 g
Catalog number B20521.06
also known as B20521-06
Price (JPY)
-
Request A Quote
Quantity:
5 g
Request bulk or custom format
Chemical Identifiers
CAS4457-72-1
IUPAC Name1,5-dibromo-3-methylpentane
Molecular FormulaC6H12Br2
InChI KeyYDPZWUMQKMLLHC-UHFFFAOYSA-N
SMILESCC(CCBr)CCBr
View more
SpecificationsSpecification SheetSpecification Sheet
Refractive Index1.5065-1.5105 @ 20?C
FormLiquid
Appearance (Color)Clear colorless to pale yellow
Assay (GC)≥98.0%
1,5-Dibromo-3-methylpentane is used as a chemical reagent, pharmaceutical intermediate, pharmaceutical research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,5-Dibromo-3-methylpentane is used as a chemical reagent, pharmaceutical intermediate, pharmaceutical research.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents, strong bases.
RUO – Research Use Only

General References:

  1. NJ Leonard.; ZW Wicks. 1, 5-Dibromo-3-methylpentane. J. Am. Chem. Soc. 194668 (11), 2402-2404.
  2. CR Johnson.; D McCants. The Stereochemistry of Oxidation at Sulfur. Oxidation of 4-Substituted Thianes to Sulfoxides1. J. Am. Chem. Soc. 196587 (5), 1109-1114.