Cyclooctanone, 98%
Cyclooctanone, 98%
Cyclooctanone, 98%
Thermo Scientific Chemicals

Cyclooctanone, 98%

CAS: 502-49-8 | C8H14O | 126.199 g/mol
Quantity:
25 g
100 g
500 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number B20911.14
also known as B20911-14
Price (JPY)
-
Request A Quote
Quantity:
25 g
Request bulk or custom format
Chemical Identifiers
CAS502-49-8
IUPAC Namecyclooctanone
Molecular FormulaC8H14O
InChI KeyIIRFCWANHMSDCG-UHFFFAOYSA-N
SMILESO=C1CCCCCCC1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to pale cream
Assay (GC)≥97.5%
FormCrystalline or fused solid
Identification (FTIR)Conforms
Cyclooctanone is a aliphatic cycle that exhibited inhibitory activity towards aldosterone synthase, a promising therapeutic target for the treatment of cardiovascular diseases related to abnormally high aldosterone level. It is also used in the synthesis of 14-membered lactones.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Soluble in water at 20°C 15g/L. Soluble in acetone, alcohol, chloroform, methanol and benzene.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Keep ignition sources away.
RUO – Research Use Only

General References:

  1. T. Maris, et al. Investigations of the Phase Transitions in Thiourea Inclusion Compounds with Cycloheptane, Cyclooctane, and Cyclooctanone.Chem. Mater.,2001,13(8), 2483-2492.
  2. Jon G. Moehlmann and J. D. McDonald. Vibrational energy distribution of chemically activated cyclooctanone.J. Chem. Phys.,1973,59(12), 6683.