Di-n-propyl ether, 99%
Di-n-propyl ether, 99%
Di-n-propyl ether, 99%
Thermo Scientific Chemicals

Di-n-propyl ether, 99%

CAS: 111-43-3 | C6H14O | 102.18 g/mol
Quantity:
25 g
100 g
Catalog number B21423.14
also known as B21423-14
Price (JPY)
-
Quantity:
25 g
Request bulk or custom format
Chemical Identifiers
CAS111-43-3
IUPAC Name1-propoxypropane
Molecular FormulaC6H14O
InChI KeyPOLCUAVZOMRGSN-UHFFFAOYSA-N
SMILESCCCOCCC
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
FormLiquid
Assay (GC)≥98.5%
Identification (FTIR)Conforms
Refractive Index1.3790-1.3820 @ 20?C
Di-n-propyl ether, is used as a solvent and intermediate for organic chemical synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Di-n-propyl ether, is used as a solvent and intermediate for organic chemical synthesis.

Solubility
It is slightly soluble in water 3 g/L (20°C), soluble in alcohol and ether.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents, strong acids, halogens.
RUO – Research Use Only

General References:

  1. AC Lloyd.; KR Darnall.; AM Winer.; JN Pitts. Relative rate constants for the reactions of OH radicals with isopropyl alcohol, diethyl and DI-n-propyl ether at 305±2 K. Chemical Physics Letters. 197642 (2), 205-209.
  2. L Wang.; GC Benson.; BCY Lu. Excess enthalpies for (di-n-propyl ether+ n-alkane) at 298.15 K. The Journal of Chemical Thermodynamics. 198820 (8), 975-979.