Indane, 95%
Indane, 95%
Indane, 95%
Thermo Scientific Chemicals

Indane, 95%

CAS: 496-11-7 | C9H10 | 118.179 g/mol
Quantity:
100 mL
500 mL
2500 mL
Catalog number B21442.0F
also known as B21442-0F
Price (JPY)
-
Quantity:
2500 mL
Request bulk or custom format
Chemical Identifiers
CAS496-11-7
IUPAC Name2,3-dihydro-1H-indene
Molecular FormulaC9H10
InChI KeyPQNFLJBBNBOBRQ-UHFFFAOYSA-N
SMILESC1CC2=CC=CC=C2C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to yellow
FormLiquid
Assay (unspecified)>94.0%
Refractive Index1.5340-1.5400 @ 20?C
Indane, is used as a catalytic agent, Petrochemical additive, used in organic synthesis. It is also used as pharmaceutical and chemical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Indane, is used as a catalytic agent, Petrochemical additive, used in organic synthesis. It is also used as pharmaceutical and chemical intermediate.

Solubility
Soluble in ethanol, diethyl ether, chloroform (slightly), and ether. Insoluble in water.

Notes
Keep away from acids, acid chlorides, acid anhydrides, strong oxidizing agents, carbon dioxide. Keep away from strong oxidizing agents.
RUO – Research Use Only

General References:

  1. A Ramazani.; A Bodaghi. One-pot, four-component synthesis of dialkyl [indane-1, 3-dione-2-ylidene] alkoxysuccinates. Tetrahedron Letters. 200041 (4), 567-568.
  2. MP Cava.; RL Litle.; DR Napier. Condensed cyclobutane aromatic systems. V. The synthesis of some α-diazoindanones: ring contraction in the indane series. J. Am. Chem. Soc. 195880 (9), 2257-2263.