3,4-Dihydroxybenzhydrazide, 97%
3,4-Dihydroxybenzhydrazide, 97%
3,4-Dihydroxybenzhydrazide, 97%
3,4-Dihydroxybenzhydrazide, 97%
Thermo Scientific Chemicals

3,4-Dihydroxybenzhydrazide, 97%

CAS: 39635-11-5 | C7H8N2O3 | 168.152 g/mol
Quantity:
5 g
25 g
Catalog number B21746.14
also known as B21746-14
Price (JPY)
-
Request A Quote
Quantity:
25 g
Request bulk or custom format
Chemical Identifiers
CAS39635-11-5
IUPAC Name3,4-dihydroxybenzohydrazide
Molecular FormulaC7H8N2O3
InChI KeyWGXWEXNJRZMIPT-UHFFFAOYSA-N
SMILESNNC(=O)C1=CC=C(O)C(O)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to pale brown
FormPowder
Assay (HPLC)≥96.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only