DL-Tropic acid, 98%
DL-Tropic acid, 98%
DL-Tropic acid, 98%
Thermo Scientific Chemicals

DL-Tropic acid, 98%

CAS: 552-63-6 | C9H10O3 | 166.18 g/mol
Quantity:
10 g
50 g
Catalog number B22040.09
also known as B22040-09
Price (JPY)
-
Quantity:
10 g
Request bulk or custom format
Chemical Identifiers
CAS552-63-6
IUPAC Name3-hydroxy-2-phenylpropanoic acid
Molecular FormulaC9H10O3
InChI KeyJACRWUWPXAESPB-UHFFFAOYNA-N
SMILESOCC(C(O)=O)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
FormCrystals or powder or crystalline powder
Assay (Aqueous acid-base Titration)≥97.5 to ≤102.5%
Assay (Silylated GC)≥97.5%
Identification (FTIR)Conforms
View more
It is a laboratory reagent used in the chemical synthesis of atropine and hyoscyamine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is a laboratory reagent used in the chemical synthesis of atropine and hyoscyamine.

Solubility
Soluble in water (20 g/L) at 20°C, ethanol, ether, alcohol, and methanol (0.1 g/ml).

Notes
Store in cool, dry place in tightly closed containers. Store away from strong oxidizing agents.
RUO – Research Use Only

General References:

  1. Edward Leete. The Biogenesis of Tropic Acid and Related Studies on the Alkaloids of Datura stramonium. J. Am. Chem. Soc. 1960, 82 (3), 612-614.
  2. Marc-André Fliniaux; Françoise Manceau; Annie Jacquin-Dubreuil. Simultaneous analysis of l-hyoscyamine, l-scopolamine and dl-tropic acid in plant material by reversed-phase high-performance liquid chromatography. Journal of Chromatography A. 1993, 644 (1), 193-197.