4-n-Pentylcyclohexanone, 98+%
4-n-Pentylcyclohexanone, 98+%
4-n-Pentylcyclohexanone, 98+%
4-n-Pentylcyclohexanone, 98+%
Thermo Scientific Chemicals

4-n-Pentylcyclohexanone, 98+%

CAS: 61203-83-6 | C11H20O | 168.28 g/mol
Quantity:
10 g
50 g
Catalog number B22088.18
also known as B22088-18
Price (JPY)
-
Request A Quote
Quantity:
50 g
Request bulk or custom format
Chemical Identifiers
CAS61203-83-6
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to pale yellow
FormLiquid
Assay (GC)≥96.0%
Refractive Index1.4550-1.4590 @ 20°C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only