Thermo Scientific Chemicals

Benzyl 4-bromophenyl ketone, 98%, Thermo Scientific Chemicals

Catalog number: B22159.03
1 g, Each
Thermo Scientific Chemicals

Benzyl 4-bromophenyl ketone, 98%, Thermo Scientific Chemicals

Catalog number: B22159.03
1 g, Each
Quantity
Catalog number: B22159.03
also known as B22159-03
Price (USD)
Price: 60.30
Online price: 51.65
Your price:
Quantity
-

Chemical Identifiers

CAS
2001-29-8
IUPAC Name
1-(4-bromophenyl)-2-phenylethan-1-one
Molecular Formula
C14H11BrO
InChI Key
MOSIKPSTRPODHQ-UHFFFAOYSA-N
SMILES
BrC1=CC=C(C=C1)C(=O)CC1=CC=CC=C1
Assay (GC)
≥97.5%
Melting Point (clear melt)
107.5-113.5?C
Appearance (Color)
White to pale yellow
Form
Crystals or powder or crystalline powder

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text