4,4'-Diaminobenzanilide, 98%
4,4'-Diaminobenzanilide, 98%
4,4'-Diaminobenzanilide, 98%
Thermo Scientific Chemicals

4,4'-Diaminobenzanilide, 98%

CAS: 785-30-8 | C13H13N3O | 227.27 g/mol
Quantity:
10 g
50 g
Catalog number B22419.18
also known as B22419-18
Price (JPY)
-
Quantity:
50 g
Request bulk or custom format
Chemical Identifiers
CAS785-30-8
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White or grey to pale brown
FormPowder or crystalline powder
Assay (Non-aqueous acid-base Titration)≥97.5 to ≤102.5%
Assay (HPLC)≥97.5%
Melting Point (clear melt)203.0-209.0°C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only