Sodium methanesulfonate, 98%
Sodium methanesulfonate, 98%
Sodium methanesulfonate, 98%
Thermo Scientific Chemicals

Sodium methanesulfonate, 98%

CAS: 2386-57-4 | CH3NaO3S | 118.082 g/mol
Quantity:
5 g
25 g
Catalog number B22439.06
also known as B22439-06
Price (JPY)
-
Quantity:
5 g
Request bulk or custom format
Chemical Identifiers
CAS2386-57-4
IUPAC Namemethanesulfonic acid sodium
Molecular FormulaCH4NaO3S
InChI KeyDWNWZFAKFWMPTI-UHFFFAOYSA-N
SMILES[Na].CS(O)(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
FormCrystals or powder or crystalline powder
Assay (ex Sulphated Ash)≥97.5 to ≤102.5%
Loss on Drying≤1.0% (200?C/constant wt)
Sulfated Ash58.95-61.35%
Sodium methanesulfonate is used as a non-steroidal anti-inflammatory drug.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Sodium methanesulfonate is used as a non-steroidal anti-inflammatory drug.

Solubility
Soluble in water, alcohol, ether, hexane, benzene, methylcyclopentane, toluene, o-chlorotoluene and ethyl disulfide.

Notes
Hygroscopic. Incompatible with strong oxidizing agents.
RUO – Research Use Only

General References:

  1. Rodriguez, E. S.; Poynter, S.; Curran, M.; Haddad, P. R.; Shellie, R. A.; Nesterenko, P. N.; Paull, B. Capillary ion chromatography with on-column focusing for ultra-trace analysis of methanesulfonate and inorganic anions in limited volume Antarctic ice core samples. J. Chromatogr. A 2015, 1409, 182-188.
  2. Wallace, M.; Iggo, J. A.; Adams, D. J. Using solution state NMR spectroscopy to probe NMR invisible gelators. Soft matter 2015, 11 (39), 7739-7747.