1,4-Diiodooctafluorobutane, 97%, stab. with copper
1,4-Diiodooctafluorobutane, 97%, stab. with copper
1,4-Diiodooctafluorobutane, 97%, stab. with copper
Thermo Scientific Chemicals

1,4-Diiodooctafluorobutane, 97%, stab. with copper

CAS: 375-50-8 | C4F8I2 | 453.84 g/mol
Quantity:
5 g
25 g
Catalog number B22660.14
also known as B22660-14
Price (JPY)
-
Quantity:
25 g
Request bulk or custom format
Chemical Identifiers
CAS375-50-8
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear, colorless to green/blue or pink
FormLiquid
Assay (GC)>96.0%
Refractive Index1.4269-1.4319 @ 20?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only