Thermo Scientific Chemicals

Diethyl n-propylmalonate, 99%, Thermo Scientific Chemicals

Catalog number: B23304.22
100 g, Each
Thermo Scientific Chemicals

Diethyl n-propylmalonate, 99%, Thermo Scientific Chemicals

Catalog number: B23304.22
100 g, Each
Quantity
Catalog number: B23304.22
also known as B23304-22
Price (USD)
Price: 148.00
Online price: 127.65
Your price:
Quantity
-

Chemical Identifiers

CAS
2163-48-6
IUPAC Name
1,3-diethyl 2-propylpropanedioate
Molecular Formula
C10H18O4
InChI Key
GRRSDGHTSMJICM-UHFFFAOYSA-N
SMILES
CCCC(C(=O)OCC)C(=O)OCC
Form
Liquid
Refractive Index
1.4190-1.4220 @ 20?C
Appearance (Color)
Clear colorless
Assay (GC)
≥98.5%

Description

Diethyl n-propylmalonate was used in the synthesis of propylacrylic acid (PAA).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Diethyl n-propylmalonate was used in the synthesis of propylacrylic acid (PAA).

Solubility
Soluble in Dichloromethane, Ethanol and Ethyl Acetate.

Notes
Store in a cool, dry conditions, in well sealed conditions. Store away from oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text