Thermo Scientific Chemicals

4-Formylmorpholine, 99%, Thermo Scientific Chemicals

Catalog number: B23315.36
500 g, Each
Thermo Scientific Chemicals

4-Formylmorpholine, 99%, Thermo Scientific Chemicals

Catalog number: B23315.36
500 g, Each
Quantity
Catalog number: B23315.36
also known as B23315-36
Price (USD)
Price: 162.00
Online price: 138.65
Your price:
Quantity
-

Chemical Identifiers

CAS
4394-85-8
IUPAC Name
morpholine-4-carbaldehyde
Molecular Formula
C5H9NO2
InChI Key
LCEDQNDDFOCWGG-UHFFFAOYSA-N
SMILES
O=CN1CCOCC1
Appearance (Color)
Clear colorless
Refractive Index
1.4850-1.4880 @ 20?C
Form
Liquid
Identification (FTIR)
Conforms
Assay (GC)
≥98.5%

Description

4-Formylmorpholine is widely used as a solvent and extracting agent for aromatics. It is used as a scrubbing agent for acidic gases. It is employed in the preparation of adenine hydrochloride labeled with 14C, 4-(trifluoromethyl)morpholine, 3-alkyl-2-chloro-5,6-dihydrobenzaldehyde and terphenyl dialdehyde. It plays an important role in the desulfurization of natural gas, synthetic gas, flue gas and gasoline.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Formylmorpholine is widely used as a solvent and extracting agent for aromatics. It is used as a scrubbing agent for acidic gases. It is employed in the preparation of adenine hydrochloride labeled with 14C, 4-(trifluoromethyl)morpholine, 3-alkyl-2-chloro-5,6-dihydrobenzaldehyde and terphenyl dialdehyde. It plays an important role in the desulfurization of natural gas, synthetic gas, flue gas and gasoline.

Solubility
Miscible with water.

Notes
Store in a cool place. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text