3-Phenylpyridine, 95%
3-Phenylpyridine, 95%
3-Phenylpyridine, 95%
Thermo Scientific Chemicals

3-Phenylpyridine, 95%

CAS: 1008-88-4 | C11H9N | 155.2 g/mol
Quantity:
5 g
25 g
Catalog number B23930.14
also known as B23930-14
Price (JPY)
-
Quantity:
25 g
Request bulk or custom format
Chemical Identifiers
CAS1008-88-4
IUPAC Name3-phenylpyridine
Molecular FormulaC11H9N
InChI KeyHJKGBRPNSJADMB-UHFFFAOYSA-N
SMILESC1=CC=C(C=C1)C1=CC=CN=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to yellow
Refractive Index1.6135-1.6195 @ 20?C
Assay (GC)≥94.0%
FormLiquid
3-Phenylpyridine is a useful synthetic intermediate. It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Phenylpyridine is a useful synthetic intermediate. It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
Soluble in chloroform, dichloromethane and ethyl acetate.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible material is strong oxidizing agents.
RUO – Research Use Only

General References:

  1. Thomas L. Perry; Karen Jones; Shirley Hansen; Richard A. Wall. 2-Phenylpyridine and 3-phenylpyridine, constituents of tea, are unlikely to cause idiopathic Parkinson's disease. Journal of the Neurological Sciences. 1988, 85 (3), 309-317.
  2. J. Kubin; A.C. Testa. Excited state protonation of phenylpyridines. Journal of Photochemistry and Photobiology A: Chemistry. 1994, 83 (2), 91-96.