(1-Butyl)triethylammonium bromide, 97%
(1-Butyl)triethylammonium bromide, 97%
(1-Butyl)triethylammonium bromide, 97%
Thermo Scientific Chemicals

(1-Butyl)triethylammonium bromide, 97%

CAS: 13028-69-8 | C10H24BrN | 238.213 g/mol
Quantity:
25 g
100 g
Catalog number B24128.14
also known as B24128-14
Price (JPY)
-
Quantity:
25 g
Request bulk or custom format
Chemical Identifiers
CAS13028-69-8
IUPAC Namebutyltriethylazanium
Molecular FormulaC10H24N
InChI KeyMPDDDPYHTMZBMG-UHFFFAOYSA-N
SMILESCCCC[N+](CC)(CC)CC
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (Titration ex Bromide)≥96.0 to ≤104.0%
Appearance (Color)White
FormCrystals or powder or crystalline powder
Water Content (Karl Fischer Titration)≤1.5%

(1-Butyl)triethylammonium bromide is an important organic intermediate. It can be used in agrochemical, pharmaceutical & dye stuff field.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(1-Butyl)triethylammonium bromide is an important organic intermediate. It can be used in agrochemical, pharmaceutical & dye stuff field.

Solubility
Soluble in water.

Notes
Hygroscopic. Store away from oxidizing agents, water/ moisture. Store under inert gas.
RUO – Research Use Only

General References:

  1. Joseph Paul Jayachandran; Thayikkannu Balakrishnan; Maw-Ling Wang. Phase-transfer-catalyzed Darzen's condensation of chloroacetonitrile with cyclohexanone using aqueous sodium hydroxide and a new phase transfer catalyst. Journal of Molecular Catalysis A: Chemical. 2000, 152 (1-2), 91-98.