2,2-Dimethylsuccinic acid, 99%
2,2-Dimethylsuccinic acid, 99%
2,2-Dimethylsuccinic acid, 99%
Thermo Scientific Chemicals

2,2-Dimethylsuccinic acid, 99%

CAS: 597-43-3 | C6H10O4 | 146.14 g/mol
Quantity:
5 g
25 g
Catalog number B24394.14
also known as B24394-14
Price (JPY)
-
Quantity:
25 g
Request bulk or custom format
Chemical Identifiers
CAS597-43-3
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Assay (Aqueous acid-base Titration)≥98.5 to ≤101.5%
Melting Point (clear melt)138.0-144.0?C
Assay (Silylated GC)≥98.5%
FormCrystals or powder or crystalline powder
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only