2-Chloro-4,6-dimethoxy-1,3,5-triazine, 98%
2-Chloro-4,6-dimethoxy-1,3,5-triazine, 98%
2-Chloro-4,6-dimethoxy-1,3,5-triazine, 98%
2-Chloro-4,6-dimethoxy-1,3,5-triazine, 98%
Thermo Scientific Chemicals

2-Chloro-4,6-dimethoxy-1,3,5-triazine, 98%

CAS: 3140-73-6 | C5H6ClN3O2 | 175.57 g/mol
Quantity:
5 g
25 g
100 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number B24474.22
also known as B24474-22
Price (JPY)
-
Request A Quote
Quantity:
100 g
Request bulk or custom format
Chemical Identifiers
CAS3140-73-6
SpecificationsSpecification SheetSpecification Sheet
Assay (unspecified)≥97.5%
Appearance (Color)White
FormPowder
2-Chloro-4,6-dimethoxy-1,3,5-triazine is used as a peptide coupling agent for the purification of peptides. It is used in the synthesis of 4-(4,6-dimethoxy-1,3,5-triazin-2-yl)-4-methylmorpholinium chloride through coupling with N-methylmorpholine in tetrahydrofuran. Further, it plays an important role in the production of bis(4,6-dimethoxy-1,3,5-triazin-2-yl) ether, through reaction with 2-hydroxy-4,6-dimethoxy-1,3,5-triazine. In addition to this, it is involved in the preparation of 2-(4,6-dimethoxy-1,3,5-triazinyl)trialkylammonium salts through reaction with various tertiary amines.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Chloro-4,6-dimethoxy-1,3,5-triazine is used as a peptide coupling agent for the purification of peptides. It is used in the synthesis of 4-(4,6-dimethoxy-1,3,5-triazin-2-yl)-4-methylmorpholinium chloride through coupling with N-methylmorpholine in tetrahydrofuran. Further, it plays an important role in the production of bis(4,6-dimethoxy-1,3,5-triazin-2-yl) ether, through reaction with 2-hydroxy-4,6-dimethoxy-1,3,5-triazine. In addition to this, it is involved in the preparation of 2-(4,6-dimethoxy-1,3,5-triazinyl)trialkylammonium salts through reaction with various tertiary amines.

Solubility
Soluble in water, chloroform and methanol.

Notes
Incompatible with strong oxidizing agents.
RUO – Research Use Only

General References:

  1. Reagent for activation of carboxylic acids to nucleophiles: J. Org. Chem., 63, 4248 (1998). Peptide coupling reagent: Synthesis, 917 (1987). With N-methylmorpholine forms the quaternary chloride (DMTMM) which is a valuable low-cost coupling agent in solid-phase peptide synthesis: Synlett, 275 (2000). For a reexamination of the use of the reagent in peptide coupling, see: Tetrahedron Lett., 42, 4161 (2002); see also Appendix 6.
  2. Similarly, carboxylic acids are converted to Weinreb amides by reaction with N,O-dialkyl hydroxylamines: J. Org. Chem., 66, 2534 (2001).
  3. Can also be used for the esterification of carboxylic acids: Synthesis, 593 (1999). The intermediate active esters can be hydrogenolyzed, providing a simple method for reduction of acids to aldehydes; at higher pressures, further reduction to the alcohol can be effected: J. Org. Chem., 64, 8962 (1999).
  4. Iranpoor, N.; Panahi, F.; Jamedi, F. Nickel-catalyzed one-pot synthesis of biaryls from phenols and arylboronic acids via C-O activation using TCT reagent. J. Organomet. Chem. 2015, 781, 6-10.
  5. Duangkamol, C.; Jaita, S.; Wangngae, S.; Phakhodee, W.; Pattarawarapan, M. Catalytic role of PPh3 and its polymer bound analog in the amidation of carboxylic acids mediated by 2,4,6-trichloro-1,3,5-triazine. Tetrahedron Lett. 2015, 56 (35), 4997-5001.