Thermo Scientific Chemicals

2-Bromo-3,3,3-trifluoro-1-propene, 97%, Thermo Scientific Chemicals

Catalog number: B24614.06
5 g, Each
Thermo Scientific Chemicals

2-Bromo-3,3,3-trifluoro-1-propene, 97%, Thermo Scientific Chemicals

Catalog number: B24614.06
5 g, Each
Quantity
Catalog number: B24614.06
also known as B24614-06
Price (USD)
Quantity
-

Chemical Identifiers

CAS
1514-82-5
Appearance (Color)
Clear colorless
Assay (GC)
≥96.0%
Comment
Specification differs for U.S. and non-U.S. material where indicated
Refractive Index
1.3515-1.3565 @ 20°C (non-U.S. specification)
Form
Liquid

Description

It is used as intermediate in production of organ fluorine compounds & as a monomer in fluoropolymer production. It is used as a trifluoromethylated synthetic building block for the synthesis of trifluoromethyl-substituted molecules.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as intermediate in production of organ fluorine compounds & as a monomer in fluoropolymer production. It is used as a trifluoromethylated synthetic building block for the synthesis of trifluoromethyl-substituted molecules.

Solubility
Soluble in most organic solvents, immiscible with water.

Notes
Store in a cool, dry place in tightly closed container. Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text