Cyclohexylurea, 98%
Cyclohexylurea, 98%
Cyclohexylurea, 98%
Thermo Scientific Chemicals

Cyclohexylurea, 98%

CAS: 698-90-8 | C7H14N2O | 142.202 g/mol
Quantity:
25 g
100 g
Catalog number B24915.14
also known as B24915-14
Price (JPY)
-
Request A Quote
Quantity:
25 g
Request bulk or custom format
Chemical Identifiers
CAS698-90-8
IUPAC Namecyclohexylurea
Molecular FormulaC7H14N2O
InChI KeyWUESWDIHTKHGQA-UHFFFAOYSA-N
SMILESNC(=O)NC1CCCCC1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
FormCrystals or powder or crystalline powder
Assay (GC)≥97.5%
Melting Point (clear melt)191.0-197.0?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only