Specifications
CAS306935-88-6
Chemical Name or Material3-Bromo-4-fluorobiphenyl
DOT InformationTransport Hazard Class: 9; Packing Group: II; Proper Shipping Name: POLYHALOGENATED BIPHENYLS, LIQUID
Formula Weight251.1
Health Hazard 1H315-H319-H335
View more